missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media
Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
Supplier: BD 221196
Description
This medium is capable of supporting good growth of a great variety of fastidious organisms, of both pathogenic and nonpathogenic species.
- Neutralize antibacterial activity of mercurial compounds
- Before use ∽30% of media should oxidized
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
| CE, AOAC, BAM, BS12, COMPF, EP, USDA, USP | |
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L | |
| Tube |
| Light Beige | |
| Liquid | |
| Thioglycollate (Fluid) | |
| Sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles | |
| 100 x 8 mL |
For in vitro diagnostic use.