missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Middlebrook OADC Enrichment

Catalog No. B11886
Change view
Click to view available options
Quantity:
500 mL
10 x 20 mL
Packaging Type:
Bottle
Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B11886 10 x 20 mL Tube
B12351 500 mL Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11886 Supplier BD Supplier No. 211886

Used as a growth supplement for microbiology research. BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.

  • Dehydrated format
  • Culture media
  • DNR (Latex) free

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Middlebrook OADC Enrichment
Form Viscous
For Use With (Application) Dehydrated culture media
Packaging Type Tube
Quantity 10 x 20 mL