missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Reagents

Offers superior detection time in a user-friendly, fluorescent test format

$195.18 - $317.68

Specifications

Certifications/Compliance CE
Content And Storage 2 to 25°C
For Use With (Application) For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
Quantity 6 Vials
View More Specs

Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
B4345114
View Documents
BD
245114
Pack of 6 Each for $195.18
Only null left
 
B4345116
View Documents
BD
245116
Pack of 6 Each for $317.68
Only null left
 
Description

Description

  • Creates an environment suitable for rapid mycobacterial growth
  • Recommended for testing sputum, stool specimens, body fluids and tissue
  • The tube uses fluorometric technology to aid in the reading of results
  • Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
  • The Mycobacteria can be removed from the tube for further testing
  • To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
  • Tubes fit into standard procedures for decontamination and waste disposal

Specifications

Specifications

CE
2 to 25°C
For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
6 Vials
Videos
SDS
Documents

Documents