missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ IsoVitaleX™ Enrichment

BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.

$329.39 - $329.39

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive IsoVitaleX™ Enrichment
Form Lyophilized
For Use With (Application) Dehydrated culture media
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11876
View Documents
BD
211876
5 x 10 mL N/A
Only null left
 
Description

Description

  • GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
  • Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
  • Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
Specifications

Specifications

ISO/DIS, 11014
IsoVitaleX™ Enrichment
Dehydrated culture media
2°C to 8°C
Lyophilized
Vial
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'