missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gelysate™ Peptone
GREENER_CHOICE

Catalog No. B11870
Change view
Click to view available options
Quantity:
454 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11870 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11870 Supplier Gibco™ Supplier No. 211870

Gelysate Peptone is a pancreatic digest of gelatin, derived from porcine animal origin (AO) sources. Gelatin hydrolysate is high in proline residues. Gelysate Peptone is deficient in carbohydrates and is characterized by low cystine, methionine, and tryptophan content.

Gelysate Peptone is a pancreatic digest of gelatin, derived from porcine animal origin (AO) sources. Gelatin hydrolysate is high in proline residues. Gelysate Peptone is deficient in carbohydrates and is characterized by low cystine, methionine, and tryptophan content.

Specifications

Certifications/Compliance LUO
Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Additive Peptone
Form Solid
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging Type Bottle
Product Line Gelysate
Quantity 454 g