missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Saccharose

Catalog No. DF0176179
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0176179 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0176179 Supplier BD Supplier No. 217610
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-saccharose, sucrose

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Saccharose
Form Powder
For Use With (Application) Dehydrated culture media
Packaging Type Bottle
Quantity 500 g