missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase Concentrate
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Concentration | 1000000U/mL |
| Additive | Penicillinase |
| Form | Liquid |
| For Use With (Application) | Formulation of culture media for the cultivation of heterotrophic microorganisms |
| Packaging Type | Bottle |
| Quantity | 100 mL |