missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Penicillinase Concentrate

Catalog No. B11898
Change view
Click to view available options
Quantity:
20 mL
100 mL
Packaging Type:
Bottle
Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B11898 20 mL Tube
B11899 100 mL Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11898 Supplier BD Supplier No. 211898
Only null left

For cultivation of heterotrophic microorganisms

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 10000000U/mL
Additive Penicillinase
Form Liquid
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging Type Tube
Quantity 20 mL