missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Casamino Acids, vitamin assay
GREENER_CHOICE

Catalog No. p-4886993
Change view
Click to view available options
Quantity:
100 g
500 g
For Use With (Application):
Vitamin assay
Grade:
Vitamin Assay
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application) Grade
DF0288156 100 g Vitamin assay Vitamin Assay
DF0288174 500 g Vitamin assay Vitamin Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0288156 Supplier Gibco™ Supplier No. 228820
Only null left

Excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production. Gibco™ Bacto™ Casamino Acids is an animal-origin, acid hydrolysis of casein having low sodium chloride and iron concentrations.

  • Prepared according to the method described by Mueller and Miller
  • Supplemented with growth factors, cystine, maltose and iron and inorganic salts
  • Successful in fermentation for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
  • Support the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus and Escherichia
  • Dehydrated format with Peptones / Hydrolysates
  • DNR (Latex) free

Specifications

Certifications/Compliance AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Additive Casamino Acid
Form Dehydrated Powder
Grade Vitamin Assay
For Use With (Application) Vitamin assay
Packaging Type Bottle
Product Line Difco
Quantity 100 g