missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ Casamino Acids, vitamin assay

Description
- Prepared according to the method described by Mueller and Miller
- Supplemented with growth factors, cystine, maltose and iron and inorganic salts
- Successful in fermentation for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
- Support the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus and Escherichia
- Dehydrated format with Peptones / Hydrolysates
- DNR (Latex) free
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014 |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Additive | Casamino Acid |
| Form | Dehydrated Powder |
| Grade | Vitamin Assay |
| For Use With (Application) | Vitamin assay |
| Packaging Type | Bottle |
| Product Line | Difco |
| Quantity | 100 g |