missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptone
GREENER_CHOICE

Catalog No. DF0123173
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0123173 500 g
DF0123084 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0123173 Supplier Gibco™ Supplier No. 211705

missing translation for 'validMainframeMsg'

An animal origin (AO) pancreatic digest of casein.

  • Bacto Tryptone is an animal origin (AO) pancreatic digest of casein
  • It is included in standard methods manual applications and is listed in the Reagent section of the United States Pharmacopeia as meeting the specifications for a pancreatic digest of casein, a component in many of the media listed
  • The nutritional profile of this supplement is unique in that it provides a range of nutrients
  • Customers have used this supplement in fermentation applications and shown that it successfully supports the growth of many bacterial species during the production of animal health vaccines
  • This includes Escherichia coli, and Fusobacterium and Agrobacterium species when used together with Bacto Yeast Extract
  • Store in cool, dry conditions in well-sealed containers at 2° to 25°C

Features:

  • Pancreatic digest of casein
  • Developed while investigating a peptone particularly suitable for the elaboration of indole by bacteria
  • Animal origin nutritional supplement

Applications of Bacto Tryptone include:

  • Works well in fermentation applications
  • Use with common organisms such as Escherichia coli, as well as uncommon organisms such as the diatom Nitzschia laevis

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Dehydrated
Product Type Bacterial Media
Format Bottle
For Use With (Application) Dehydrated culture media
Product Line Bacto
Quantity 500 g
Source Animal Origin