missing translation for 'onlineSavingsMsg'
Learn More

BCL2L2 293T Cell Overexpression Lysate (Denatured), Abnova

Catalog No. 89000292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89000292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89000292 Supplier Abnova Corporation Supplier No. H00000599T01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Western Blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description BCL2L2 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_004050
Molecular Weight (g/mol) 21.34
pH Range 6.8
Plasmid pCMV-BCL2L2 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-BCL2L2 antibody (H00000599-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less