missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCL2A1 Recombinant Rabbit Monoclonal Antibody (23GB4820)

Catalog No. MA552757
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552757 20 μL
MA552756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552757 Supplier Invitrogen™ Supplier No. MA552757
Only null left

Rabbit Recombinant Monoclonal Antibody

BCL2A1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Bcl-2 related Protein A1 is a member of the BCL-2 protein family. The proteins of this family form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities such as embryonic development, homeostasis and tumorigenesis. The protein encoded by this gene is able to reduce the release of pro-apoptotic cytochrome c from mitochondria and block caspase activation. This gene is a direct transcription target of NF-kappa B in response to inflammatory mediators, and has been shown to be up-regulated by different extracellular signals, such as granulocyte-macrophage colony-stimulating factor (GM-CSF), CD40, phorbol ester and inflammatory cytokine TNF and IL-1, which suggests a cytoprotective function that is essential for lymphocyte activation as well as cell survival.

Specifications

Antigen BCL2A1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4820
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCL2A1
Gene Accession No. Q16548
Gene Alias A1; A1-A; ACC1; ACC-1; ACC2; ACC-2; B cell leukemia/lymphoma 2 related protein A1a; BB218357; B-cell leukemia/lymphoma 2 related protein A1; B-cell leukemia/lymphoma 2 related protein A1a; B-cell leukemia/lymphoma 2 related protein A1d; BCL2 related protein A1; Bcl2a1; Bcl2a1a; Bcl2a1d; BCL2L5; bcl2-L-5; Bcl-2-like protein 5; bcl-2-related protein A1; BCL2-related protein A1; BCL2-related protein A1d; Bfl1; Bfl-1; GRS; GRS protein; HBPA1; hematopoietic BCL2-related protein A1; hemopoietic-specific early response protein; Protein BFL-1; Protein GRS
Gene Symbols BCL2A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCL2A1 (1-60AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 597
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less