missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Bcl-6 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1114S1
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1114S1 500 μL Concentrated
MS1114S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MS1114S1 Supplier Epredia™ Supplier No. MS1114S1
Only null left

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Bcl-6 Ab-2, Mouse Monoclonal Antibody.

Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.

Host Species: Mouse

Clone: BL6.02, same as PG-B6p

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rabbit, Rat, Sheep and Pig. Others not tested.

Epitope: aa 3-484

Immunogen: Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bcl-6 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BL6.02; same as PG-B6p
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein
Purification Method Tonsil
Quantity 500 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Bovine, Human, Pig, Rabbit, Rat, Sheep
Form Concentrated
Isotype IgG2a κ
Show More Show Less