missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCAT2 Monoclonal Antibody (7G3A11)
GREENER_CHOICE

Catalog No. FERMA548386
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548386 Supplier Invitrogen™ Supplier No. MA548386
Only null left

Mouse Monoclonal Antibody

BCAT2 Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, ELISA

This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen BCAT2
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7G3A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BCAT2
Gene Accession No. O15382
Gene Alias BCAM; BCAT(m); Bcat2; Bcat-2; Bcatm; BCT2; branched chain amino acid transaminase 2; branched chain amino acid transaminase 2, mitochondrial; branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; branched-chain-amino-acid aminotransferase, mitochondrial; Eca40; heart branched chain aminotransferase precursor encoding mitochondrial protein; mBcat; placental protein 18; PP18
Gene Symbols BCAT2
Host Species Mouse
Immunogen Purified recombinant fragment of human BCAT2 (AA: 259-392) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less