Learn More
BD BBL™ Taxo™ Blank Discs
Provides a cost effective identification and differentiation tool. BD™ BBL™ Taxo™ Blank Discs are easy-to-use discs for classical identification of microorganisms. The Taxo product line includes discs for identifying a variety of organisms.
$415.39 - $2,546.72
Specifications
For Use With (Application) Identification and differentiation of microorganisms
For Use With (Equipment) Taxo™ brand testing disc
Packaging Type BBL Blank Test Discs
Quantity 6 Pack
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Outside Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Outside Price Quantity & Availability  
B31122
View Documents Promotion Details
BD
231122
1/2 in.
Case of 6 Each for $2,546.72
Only null left
B31039
View Documents Promotion Details
BD
231039
1/4 in.
Case of 6 Each for $415.39
Only null left
Description
Description
Order Info
50 discs per vial; six vials per pack
Specifications
Specifications
Identification and differentiation of microorganisms
Taxo™ brand testing disc
BBL Blank Test Discs
6 Pack
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1825 Days