Learn More
BD BBL™ Taxo™ Blank Discs
Provides a cost effective identification and differentiation tool. BD™ BBL™ Taxo™ Blank Discs are easy-to-use discs for classical identification of microorganisms. The Taxo product line includes discs for identifying a variety of organisms.
Supplier:  BD 231122
 View more versions of this product
Catalog No. B31122
Only null left
Description
Description
Order Info
50 discs per vial; six vials per pack
Specifications
Chemical Identifiers
Identification and differentiation of microorganisms
Taxo™ brand testing disc
BBL Blank Test Discs
6 Pack
Specifications
APHA, Standard Methods for the Examination of Dairy Products
Taxo™ brand testing disc
6 Pack
12.7 mm
Identification and differentiation of microorganisms
BBL Blank Test Discs
1/2 in.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 1825 Days