missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bax Monoclonal Antibody (6A7), eBioscience™
GREENER_CHOICE

Catalog No. p-7167589
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5013581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5013581 Supplier Invitrogen™ Supplier No. 14699782
Only null left

Mouse Monoclonal Antibody

Description: The 6A7 antibody reacts with mouse, rat, monkey and human Bax (20 kDa). The 6A7 antibody shows no cross-reaction with Bcl-2 or Bcl-xL proteins. Bcl-2, Bax, and Bcl-xL are members of a protein family that regulates apoptosis; while several family members including Bcl-2 and Bcl-xL inhibit apoptosis, overexpression of Bax promotes programmed cell death. A human Bax protein peptide sequence, aa 12-24, was used as immunogen. Applications Reported: The 6A7 antibody has been reported for use in immunoblotting (WB). Applications Tested: The 6A7 antibody has been tested by Western Blot and can be used at approximately 1 μg/mL. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

BAX is a members of the Bcl-2 Family and plays an important role in regulation of apoptosis. Whereas Bcl-2 is commonly regarded as an anti-apoptotic protein, BAX is considered to have a pro-apoptotic function. Regulation of apoptosis is supposed to involve both homo- and heterodimerization of different isoforms of BAX and Bcl-2. The Bax gene encodes different isoforms including Bax alpha (21 kDa) and Bax beta (24 kDa), whereas both isoforms contain the BH1, BH2 and BH3 domains, Bax beta has a unique carboxyl terminus and does not contain a hydrophobic transmembrane domain. Bcl-2 is also expressed in different Isoforms. Bcl-2 beta differs in the 3' UTR and coding region compared to variant alpha. Bcl-2 beta is shorter (22 kDa) and has a distinct C-terminus compared to Bcl-2 alpha (26 kDa). BAX is a member of the BCL-2 family of proteins, which function as regulators of apoptosis. Overexpression of BAX functions to promote cell death. BAX can form homodimers and is also able to heterodimerize with other BCL-2 related proteins.

Specifications

Antigen Bax
Applications Western Blot
Classification Monoclonal
Clone 6A7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene BAX
Gene Accession No. Q07812, Q07813, Q63690
Gene Alias apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52
Gene Symbols BAX
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12028, 24887, 581
Target Species Human, Mouse, Rat, Monkey
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less