missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 98%, Thermo Scientific™
$244.25 - $857.05
Chemical Identifiers
| CAS | 52746-49-3 |
|---|---|
| Molecular Formula | 2NaO3S·xH2O |
| Molecular Weight (g/mol) | 536.5 |
| MDL Number | MFCD00149310 |
| InChI Key | PFNPMHXHQCVXED-UHFFFAOYSA-L |
| Synonym | 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate |
| PubChem CID | 54669730 |
| IUPAC Name | disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate |
| SMILES | C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC164050010
|
N/A | 1g | Glass bottle |
Each for $244.25
|
|
||||
|
AC164050050
|
N/A | 5g | Glass bottle |
Each for $857.05
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 52746-49-3 | |
| 536.5 | |
| PFNPMHXHQCVXED-UHFFFAOYSA-L | |
| 54669730 | |
| C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+] |
| 2NaO3S·xH2O | |
| MFCD00149310 | |
| 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate | |
| disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate |
Specifications
| 52746-49-3 | |
| 100.0 | |
| 98% | |
| MFCD00149310 | |
| PFNPMHXHQCVXED-UHFFFAOYSA-L | |
| disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate | |
| 54669730 | |
| 98% |
| 97.5 | |
| Authentic | |
| 2NaO3S·xH2O | |
| 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate | |
| C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+] | |
| 536.5 | |
| 536.5 | |
| Bathophenanthrolinedisulfonic acid, disodium salt hydrate |
Safety and Handling
EINECSNumber : 258-152-
RUO – Research Use Only