missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%

Catalog No. AC164050010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164050010 1 g Glass bottle
AC164050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC164050010 Supplier Thermo Scientific Chemicals Supplier No. 164050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52746-49-3
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.48
MDL Number MFCD00149310
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
PubChem CID 54669730
IUPAC Name disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 536.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bathophenanthrolinedisulfonic acid, disodium salt hydrate
Show More Show Less
EINECSNumber 258-152-

RUO – Research Use Only