Learn More
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol
Supplier: Thermo Scientific Chemicals B2324406
Description
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Bathophenanthrolinedisulfonic acid disodium salt hydrate | |
| >300°C | |
| MFCD00149310 | |
| 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate | |
| PCNDSIWXTYFWIA-UHFFFAOYSA-L | |
| disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate | |
| 54669730 | |
| 98% |
| 52746-49-3 | |
| C24H14N2Na2O6S2 | |
| 5 g | |
| Soluble in water. | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O | |
| 536.48 | |
| 536.48 (Anhydrous) |
Chemical Identifiers
| 52746-49-3 | |
| 536.48 | |
| PCNDSIWXTYFWIA-UHFFFAOYSA-L | |
| 54669730 |
| C24H14N2Na2O6S2 | |
| MFCD00149310 | |
| 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O |
Safety and Handling
EINECSNumber : 258-152-0
RTECSNumber : SF8433000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only