missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%

Catalog No. p-7026920
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23244MD 250 mg
AAB2324403 1 g
AAB2324406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB23244MD Supplier Thermo Scientific Chemicals Supplier No. B23244MD

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol

Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 52746-49-3
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.48
MDL Number MFCD00149310
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
PubChem CID 54669730
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 250 mg
Solubility Information Soluble in water.
IUPAC Name disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Formula Weight 536.48 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate
EINECSNumber 258-152-0
RTECSNumber SF8433000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only