missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bathocuproine, 0.1% (w/v), Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0, 0.1 |
| CAS Max % | 100.0, 0.1 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Chemical Name or Material | Bathocuproine |