missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproine, 0.1% (w/v), Ricca Chemical

Catalog No. 9054
Change view
Click to view available options
Quantity:
120 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
9054 120 mL Natural Poly Bottle
R09050001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9054 Supplier Ricca Chemical Company Supplier No. RI9054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52698-84-7 , 7732-18-5
Molecular Formula C26H18N2Na2O6S2
Molecular Weight (g/mol) 564.54
MDL Number MFCD00011679,MFCD03012947,MFCD00149974
InChI Key RNGKZLRAVYPLJC-UHFFFAOYSA-L
Synonym disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
PubChem CID 15678335
IUPAC Name disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
SMILES [Na+].[Na+].CC1=C(C(C2=CC=CC=C2)=C2C=CC3=C(C4=CC=CC=C4)C(=C(C)N=C3C2=N1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 120 mL
Concentration or Composition Notes 0.1% (w/v)
Chemical Name or Material Bathocuproine