missing translation for 'onlineSavingsMsg'
Learn More

Basic Violet 1, TCI America™

Catalog No. G017725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G017725G Supplier TCI America Supplier No. G017725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory crystal that is soluble in alcohol and chloroform and suitable for analytical applications.

Chemical Identifiers

CAS 8004-87-3
MDL Number MFCD00012568
Synonym Aniline Violet, Dahlia Violet B, Methyl Violet

Specifications

Color Dark Green
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Basic Violet 1
EINECSNumber (5)-1969
RTECSNumber GC8425000
TSCA Yes