missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BASIC™ Universal Absorbent Rolls

BASIC Universal Sorbents are economical, general purpose sorbents to clean up oils, coolants, solvents, and water-based fluids.

$201.47 - $201.47

Specifications

Color Gray
Material Polypropylene
Type Universal Sorbent
For Use With (Application) Chemical resistant sorbent for water, oil and solvent base liquids
Weight Capacity Heavyweight
View More Specs

Products 1
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
19-103-112
View Documents
Brady™
BRU152
150 ft. x 15 in.
Case of 2 Each for $201.47
Only null left
 
Description

Description

  • Universal absorbency designed to offer an economical, general purpose solution to cleaning
  • Sorbents absorb up to 20 times their weight in fluids
  • Continuous rolls are ideal for covering larger areas
  • Perforations every 30 in. make it easy to select smaller sizes when needed
  • Use against spills in assembly lines, printing, equipment or any general manufacturing areas
  • Economical single-ply construction
Specifications

Specifications

Gray
Universal Sorbent
Heavyweight
Polypropylene
Chemical resistant sorbent for water, oil and solvent base liquids
Videos
SDS
Documents

Documents