Learn More
Fisherbrand™ Basic Steel Work Bench Complete Plumbing Assembly
Description
- Plumbing assemblies include a fine control needle valve service turret with shank, a service hose, and a male quick connect fitting.
- These complete assemblies are designed specifically for use with Basic Work Bench in a laboratory environment.
- All quick connects have 316 stainless steel internal components.
- Keyed quick connect members will only couple with components of the same key which are identified by a color code.
Specifications
Specifications
| Product Type | Air Service Complete Plumbing Assembly |
| Color | Color Coded Orange |
| Material | Reinforced PVC Hose, Polished Chrome Turret |
| Includes | Male Quick Connect, System Hose and Service Turret |