missing translation for 'onlineSavingsMsg'
Learn More

Basic Fuchsin, Spectrum™ Chemical

Catalog No. 18600855
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
25 g
5 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18600855 100 g Amber Glass Bottle
18600853 5 g Amber Glass Bottle
18600854 25 g Amber Glass Bottle
18600856 1 kg Amber Glass Bottle
4 options

Catalog No. 18600855

Supplier: Spectrum Chemical Mfg Cor BA130100GM

Only null left

BA130, 569-61-9, C19H17N3·HCl

Spectrum™ Chemical Basic Fuchsin is a magenta dye that is a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin is a dark green crystal often used in the laboratory where it is employed to stain bacteria and in the fabrication of Schiff reagent. It is made up of a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin also has uses in the textile industry, and has many other names, including Basic Parafuchsin, Basic Red 9, Paramagenta Hydrochloride, and Pararosaniline Chloride.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.82
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
SMILES [H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Grade Reagent
Physical Form Crystalline Powder or Lumps