Learn More
Thermo Scientific Chemicals Basic Fuchsin, 88%, pure
CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol
$266.84 - $266.84
Chemical Identifiers
| CAS | 632-99-5 |
|---|---|
| Molecular Formula | C20H20ClN3 |
| Molecular Weight (g/mol) | 337.85 |
| MDL Number | MFCD00012569 |
| InChI Key | AXDJCCTWPBKUKL-UHFFFAOYSA-N |
| Synonym | Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride |
| PubChem CID | 12447 |
| IUPAC Name | 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
| SMILES | [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC419141000
|
Thermo Scientific Chemicals
419141000 |
100 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 632-99-5 | |
| 337.85 | |
| AXDJCCTWPBKUKL-UHFFFAOYSA-N | |
| 12447 | |
| [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 |
| C20H20ClN3 | |
| MFCD00012569 | |
| Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride | |
| 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
Specifications
| 1800 (at lambda min.) | |
| 632-99-5 | |
| 1.1 to 1.4 | |
| 83% min. (on dry substance) | |
| C20H20ClN3 | |
| 13, 765 | |
| Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride | |
| AXDJCCTWPBKUKL-UHFFFAOYSA-N | |
| [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 | |
| 337.85 | |
| 549 to 555nm (in 50:50 IMS:water) | |
| 88% | |
| Glass bottle | |
| 100 g | |
| Fine Powder |
| 250.0°C | |
| 83.0 | |
| 100.0 | |
| 88% | |
| MFCD00012569 | |
| 15, 5715 | |
| Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C) | |
| Authentic | |
| 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride | |
| 12447 | |
| 337.84 | |
| Pure | |
| Green | |
| 200°C | |
| Basic Fuchsin, 88% |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 211-189-6
RTECSNumber : CX9850000
TSCA : TSCA
RUO – Research Use Only