missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 88%, pure

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

Supplier:  Thermo Scientific Chemicals 419141000

 View more versions of this product

Catalog No. AC419141000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1800 (at lambda min.)
250.0°C
632-99-5
1.1 to 1.4
83% min. (on dry substance)
C20H20ClN3
13, 765
Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
AXDJCCTWPBKUKL-UHFFFAOYSA-N
[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
337.85
549 to 555nm (in 50:50 IMS:water)
88%
Glass bottle
100 g
Fine Powder
Basic Fuchsin
88%
83.0
100.0
88%
MFCD00012569
15, 5715
Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Authentic
4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
12447
337.84
Pure
Green
200°C

Chemical Identifiers

632-99-5
337.85
AXDJCCTWPBKUKL-UHFFFAOYSA-N
12447
[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
C20H20ClN3
MFCD00012569
Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 211-189-6

RTECSNumber : CX9850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only