missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BASIC™ Chemical Absorbent Pads

Catalog No. 19000674
Change view
Click to view available options
Capacity (English):
17 gal.
20 gal.
34 gal.
Weight Capacity:
Heavyweight
Lightweight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
19000674 17 gal. Lightweight
19000673 34 gal. Lightweight
19000672 20 gal. Heavyweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19000674 Supplier Brady™ Supplier No. BPH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal spill response for chemicals like: citric acid, hydrofluoric acid, sodium hydroxide and many other chemicals.

  • Single Ply Meltblown Polypropylene construction
  • Chemically inert, making it ideal for use with aggressive fluids
  • Unique color helps to set these absorbents apart for proper disposal
  • Perforated to help reduce waste
  • Dimpled for added durability and reduced linting

Specifications

Color Yellow
Capacity (English) 17 gal.
Material Polypropylene
Type Chemical Absorbent Pad
For Use With (Application) Absorbent for applications with hazardous chemicals
Weight Capacity Lightweight
Dimensions (L x W) 17 x 15 in.