Learn More
Brady™ BASIC™ Chemical Absorbent Pads
Ideal spill response for chemicals like: citric acid, hydrofluoric acid, sodium hydroxide and many other chemicals.
Supplier:  Brady™ BPH200
 View more versions of this product
Catalog No. 19000673
Only null left
Description
Description
  • Single Ply Meltblown Polypropylene construction
  • Chemically inert, making it ideal for use with aggressive fluids
  • Unique color helps to set these absorbents apart for proper disposal
  • Perforated to help reduce waste
  • Dimpled for added durability and reduced linting
Specifications
Chemical Identifiers
Yellow
Chemical Absorbent Pad
17 x 15 in.
Polypropylene
Absorbent for applications with hazardous chemicals
Specifications
Yellow
Polypropylene
Absorbent for applications with hazardous chemicals
17 x 15 in.
34 gal.
Chemical Absorbent Pad
Lightweight
Product Suggestions
Product Suggestions