missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium thiosulfate, pure, Thermo Scientific™
Chemical Identifiers
| CAS | 35112-53-9 |
|---|---|
| Molecular Formula | BaO3S2 |
| Molecular Weight (g/mol) | 249.46 |
| InChI Key | ONPIOWQPHWNPOQ-UHFFFAOYSA-L |
| Synonym | barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo |
| PubChem CID | 169663 |
| IUPAC Name | barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
| SMILES | [O-]S(=O)(=S)[O-].[Ba+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC453811000
|
N/A | 100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC453815000
|
N/A | 500g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 35112-53-9 | |
| 249.46 | |
| barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
| barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
| BaO3S2 | |
| ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
| 169663 | |
| [O-]S(=O)(=S)[O-].[Ba+2] |
Specifications
| 35112-53-9 | |
| BaS2O3 | |
| barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
| ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
| barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 169663 | |
| 97.5 to 102.5% | |
| Barium thiosulfate |
| BaO3S2 | |
| 15,995 | |
| Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol | |
| [O-]S(=O)(=S)[O-].[Ba+2] | |
| 249.46 | |
| 249.46 | |
| Pure |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 252-375-7
RUO – Research Use Only