missing translation for 'onlineSavingsMsg'
Learn More

Barium thiosulfate, pure, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 453811000

Quantity 100g

 View more versions of this product

Catalog No. AC453811000


Description

Description

Specifications

Chemical Identifiers

35112-53-9
249.46
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
BaO3S2
ONPIOWQPHWNPOQ-UHFFFAOYSA-L
169663
[O-]S(=O)(=S)[O-].[Ba+2]

Specifications

Barium thiosulfate
100g
BaS2O3
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
ONPIOWQPHWNPOQ-UHFFFAOYSA-L
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
169663
97.5 to 102.5%
35112-53-9
BaO3S2
15,995
Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol
[O-]S(=O)(=S)[O-].[Ba+2]
249.46
249.46
Pure
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 252-375-7

SDS
Documents

Documents

Promotions

Promotions