missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium thiosulfate, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 453815000
Specifications
| Barium thiosulfate | |
| 500g | |
| BaS2O3 | |
| barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
| ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
| barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 169663 | |
| 97.5 to 102.5% |
| 35112-53-9 | |
| BaO3S2 | |
| 15,995 | |
| Solubility in water: slightly soluble. Other solubilities: insoluble in alcohol | |
| [O-]S(=O)(=S)[O-].[Ba+2] | |
| 249.46 | |
| 249.46 | |
| Pure |
Chemical Identifiers
| 35112-53-9 | |
| 249.46 | |
| barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo | |
| barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
| BaO3S2 | |
| ONPIOWQPHWNPOQ-UHFFFAOYSA-L | |
| 169663 | |
| [O-]S(=O)(=S)[O-].[Ba+2] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 252-375-7
RUO â Research Use Only