missing translation for 'onlineSavingsMsg'
Learn More

Barium tetracyanoplatinate(II) tetrahydrate, 99+%, Thermo Scientific Chemicals

$589.62 - $589.62

Chemical Identifiers

CAS 13755-32-3
MDL Number MFCD00014183
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3946303
View Documents
Thermo Scientific Chemicals
03946303
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13755-32-3
MFCD00014183

Specifications

13755-32-3
3.05 g/mL
MFCD00014183
UN1564
508.56 (436.50 Anhydrous)
Premion™
Barium tetracyanoplatinate(II) tetrahydrate
100°C (decomposition)
Odorless
1 g
14,991
99.99%
Powder
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

EINECSNumber : 209-238-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only