accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OBaPtNNNN

CAS RN 13755-32-3

IUPAC Name: barium(2+) tetracyanoplatinumdiuide tetrahydrate
Synonyms: barium(2+) tetracyanoplatinumdiuide tetrahydrate
Molecular Weight (g/mol): 512.58
Molecular Formula: C4H12BaN4O4Pt
InChi Key: QNTHMRLOHDQAOV-UHFFFAOYSA-N
SMILES: O.O.O.O.[Ba++].[Pt--].C#N.C#N.C#N.C#N
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results