missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfide, 99.7% (metals basis)

Catalog No. p-7049086
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1284809 10 g
AA1284818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1284809 Supplier Thermo Scientific Chemicals Supplier No. 01284809

CAS: 21109-95-5 | BaS | 169.39 g/mol

Barium sulfide has different characters, so it is used in a variety of ways, such as in electronics, paint pigments, dehairing hides, flame retardant, luminous paints, and generating pure hydrogen sulfide. Barium sulfide can deviate X-rays and helps to give clearly image of the soft tissue. Barium sulfide is a precursor to other barium compounds and also used as a short wavelength emitters for electronic displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21109-95-5
Molecular Formula BaS
Molecular Weight (g/mol) 169.39
MDL Number MFCD00014192
InChI Key CJDPJFRMHVXWPT-UHFFFAOYSA-N
Synonym barium sulfide, thioxobarium
PubChem CID 88791
IUPAC Name barium(2+) sulfanediide
SMILES [S--].[Ba++]

Specifications

Melting Point 1200°C
Color Gray to Yellow
Physical Form Powder
Quantity 10 g
UN Number UN3134
Merck Index 14,995
Solubility Information Soluble in water but insoluble in alcohol.
Formula Weight 169.4
Percent Purity 99.7%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Density 4.25 g/mL
Assay (metals basis)
Chemical Name or Material Barium sulfide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
EINECSNumber 244-214-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only