accessibility menu, dialog, popup

UserName

SBa

CAS RN 21109-95-5

IUPAC Name: barium(2+) sulfanediide
Synonyms: BaS|barium sulfide|barium sulphide|bariumsulfid
Molecular Weight (g/mol): 169.39
Molecular Formula: BaS
InChi Key: CJDPJFRMHVXWPT-UHFFFAOYSA-N
SMILES: [S--].[Ba++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results