missing translation for 'onlineSavingsMsg'
Learn More

Barium Peroxide, Powder, 88%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B1053500GM

 View more versions of this product

Catalog No. 18609024

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1304-29-6
500 g
Reagent
1
88%
Amber Glass Bottle

Chemical Identifiers

1304-29-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.