missing translation for 'onlineSavingsMsg'
Learn More

Barium Peroxide, Powder, 88%, Spectrum™ Chemical

Catalog No. p-7165130
Change view
Click to view available options
Quantity:
12 kg
125 g
2.5 kg
25 g
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18609021 25 g Amber Glass Bottle
18609022 125 g Amber Glass Bottle
18609024 500 g Amber Glass Bottle
18609025 2.5 kg Amber Glass Bottle
18609023 12 kg Poly Pail
5 options

Catalog No. 18609021

Supplier: Spectrum Chemical Mfg Cor B105325GM

Only null left

Chemical Identifiers

CAS 1304-29-6

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 88%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.