missing translation for 'onlineSavingsMsg'
Learn More

Barium Perchlorate, Certified, 0.0100N ±0.0005N (0.005M), LabChem™

Catalog No. LC116452
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC116452 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC116452 Supplier LabChem Supplier No. LC116452

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical reagent grade barium perchlorate solution.

Chemical Identifiers

CAS 13465-95-7 , 7732-18-5
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.219
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Color Colorless
CAS Max % 99.83, 0.17
Physical Form Liquid
Quantity 1 L
Linear Formula Ba(ClO4)2
Solubility Information Soluble in water
Formula Weight 336.23
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0100N ±0.0005N (0.005M)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Chlorine; Barium
Density 1g/mL
Chemical Name or Material Barium Perchlorate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature