missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Perchlorate, 0.0100 N (N/100), Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0, 0.2 |
| CAS Max % | 100.0, 0.2 |
| Quantity | 1 L |
| Concentration or Composition (by Analyte or Components) | 0.01 Normal |
| Odor | Odorless |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Barium Perchlorate |
Safety and Handling
| ShelfLife | 18 months |