missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™

Catalog No. p-7071998
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 kg
500 g
Catalog No. Quantity
6000370 500 g
6000371 5 kg
2 options

Catalog No. 6000370

Supplier: Honeywell Chemicals 31128500G

Only null left

Puriss. p.a., ACS Reagent,≥99%,≥52% Ba basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate

Specifications

Quantity 500 g
Linear Formula Ba(NO3)2
UN Number 1446
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 261.34g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Barium Nitrate, ACS reagent
Hazard Statement P210-P220-P221-P261-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H302 + H332-H319
EINECSNumber 233-020-5
RTECSNumber CQ9625000