missing translation for 'onlineSavingsMsg'
Learn More

Barium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165129
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609014 25 g Amber Glass Bottle
18609015 125 g Amber Glass Bottle
18609016 500 g Amber Glass Bottle
18609017 2.5 kg Amber Glass Bottle
18609018 12 kg Poly Pail
18609019 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609014 Supplier Spectrum Chemical Mfg Cor Supplier No. B104525GM
Only null left

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
IUPAC Name barium(2+) dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

pH 5.0 to 8.0
CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.