missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, ACS, 99+%

Catalog No. p-3256909
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1219418 50 g
AA1219436 500 g
AA12194A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1219418 Supplier Thermo Scientific Chemicals Supplier No. 01219418
Only null left

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol

Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C (decomposition)
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range >99%
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name barium(2+) dinitrate
Formula Weight 261.35
Grade ACS
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.24
Chemical Name or Material Barium nitrate
Show More Show Less
Hazard Category H272-H301-H319-H332
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 233-020-5
RTECSNumber CQ9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only