Learn More
Barium manganate, tech. 90%
CAS: 7787-35-1 | BaMnO4 | 256.26 g/mol
$60.01 - $178.30
Chemical Identifiers
| CAS | 7787-35-1 |
|---|---|
| Molecular Formula | BaMnO4 |
| Molecular Weight (g/mol) | 256.26 |
| MDL Number | MFCD00011514 |
| InChI Key | ZZCNKSMCIZCVDR-UHFFFAOYSA-N |
| Synonym | barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese |
| PubChem CID | 3084030 |
| IUPAC Name | barium(2+);dioxido(dioxo)manganese |
| SMILES | [Ba++].[O-][Mn]([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1317206
|
Thermo Scientific Chemicals
L1317206 |
5 g |
Each for $60.01
|
|
|||||
|
AAL1317214
|
Thermo Scientific Chemicals
L1317214 |
25 g |
Each for $178.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7787-35-1 | |
| 256.26 | |
| ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
| 3084030 | |
| [Ba++].[O-][Mn]([O-])(=O)=O |
| BaMnO4 | |
| MFCD00011514 | |
| barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
| barium(2+);dioxido(dioxo)manganese |
Specifications
| 7787-35-1 | |
| BaMnO4 | |
| UN3087 | |
| barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
| [Ba++].[O-][Mn]([O-])(=O)=O | |
| 256.26 | |
| 256.28 | |
| Technical | |
| 4.85 |
| 5 g | |
| MFCD00011514 | |
| 14,981 | |
| ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
| barium(2+);dioxido(dioxo)manganese | |
| 3084030 | |
| 90% | |
| Moisture sensitive | |
| Barium manganate |
Safety and Handling
GHS H Statement
H272-H331-H302
May intensify fire; oxidizer.
Toxic if inhaled.
Harmful if swallowed.
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H272-H302+H332
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
EINECSNumber : 232-109-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only