CAS RN 7787-35-1
IUPAC Name:
barium(2+) dioxomanganesebis(olate)
Synonyms:
barium manganate
Molecular Weight (g/mol):
256.26
Molecular Formula:
BaMnO4
InChi Key:
ZZCNKSMCIZCVDR-UHFFFAOYSA-N
SMILES:
[Ba++].[O-][Mn]([O-])(=O)=O
1
–
1
of
1
results
1
Barium manganate, tech. 90%
CAS: 7787-35-1 Molecular Formula: BaMnO4 Molecular Weight (g/mol): 256.26 MDL Number: MFCD00011514 InChI Key: ZZCNKSMCIZCVDR-UHFFFAOYSA-N Synonym: barium manganate,ba.mno4,acmc-1bdpw,manganic acid h2mno4 , barium salt 1:1,barium 2+ ion manganate,barium 2+ ; dioxido dioxo manganese PubChem CID: 3084030 IUPAC Name: barium(2+);dioxido(dioxo)manganese SMILES: [Ba++].[O-][Mn]([O-])(=O)=O