missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium manganate, tech. 90%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 g |
| UN Number | UN3087 |
| Merck Index | 14,981 |
| Formula Weight | 256.28 |
| Percent Purity | 90% |
| Grade | Technical |
| Sensitivity | Moisture sensitive |
| Density | 4.85 |
| Chemical Name or Material | Barium manganate |
Safety and Handling
| Hazard Category | H272-H302+H332 |
| Hazard Statement | GHS H Statement H272-H331-H302 May intensify fire; oxidizer. Toxic if inhaled. Harmful if swallowed. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S. |
| EINECSNumber | 232-109-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only