missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide, Saturated Solution, LabChem™

Catalog No. LC116402
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC116402 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC116402 Supplier LabChem Supplier No. LC116402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Saturated aqueous solution for laboratory use.

Chemical Identifiers

CAS 12230-71-6 , 7732-18-5
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Color Colorless
CAS Max % 96.85, 3.15
Physical Form Liquid
Quantity 1 L
Linear Formula Ba(OH)2·8H2O
UN Number UN3266
Solubility Information Soluble in water
Formula Weight 315.47
Grade Not Available
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.03g/mL
Decomposition Information Barium
Density 1.03g/mL
Chemical Name or Material Barium Hydroxide, Saturated Solution
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Recommended Storage Room Temperature